Learn More
Selleck Chemical LLC Pentosan Polysulfate Sodium S3500-100mg
SDP
Catalog No. 503110144
Encompass_Preferred
Pentosan Polysulfate Sodium (PPS Elmiron) is an orally bioavailable semi-synthetic medication with anti-inflammatory and pro-chondrogenic properties Pentosan Polysulfate Sodium also exhibits anti-HIV-1 activity
Catalog No. 50-311-0144 Supplier Selleck Chemical LLC Supplier No. S3500100MG
Add to Cart
Add to Cart
Pentosan Polysulfate Sodium (PPS Elmiron) is an orally bioavailable semi-synthetic medication with anti-inflammatory and pro-chondrogenic properties Pentosan Polysulfate Sodium also exhibits anti-HIV-1 activity