Learn More
Cayman Chemical Pentosidine trifluoroacetate
SDP
Supplier:  Cayman Chemical 100102541 MG
Encompass_Preferred
A well-characterized natural advanced glycation end (AGE) that is often used as a biomarker for the production of all AGEs
Catalog No. 50-289-1026