Learn More
Cayman Chemical Pentosidine trifluoroacetate
SDP
Supplier:  Cayman Chemical 10010254500 UG
Encompass_Preferred
A well-characterized natural advanced glycation end (AGE) that is often used as a biomarker for the production of all AGEs
Catalog No. 50-288-7782