Learn More
Cayman Chemical PentostatIn 10mg
SDP
Catalog No. 502969676
Encompass_Preferred
A purine antimetabolite and an ADA inhibitor (Ki = 0.09 nM); reduces 2'-deoxyadenosine-induced growth inhibition of MOLT-4 lymphoblast cells (Ki = 5.6 µM); inhibits erythrocytes and ascites tumor cell ADA in an L1210 murine lymphocytic leukemia model at 0.2 mg/kg; inhibits tumor growth and increases survival rate in a CT26/NY-ESO-1 mouse xenograft model at 10 UG/mouse
Catalog No. 50-296-9676 Supplier Cayman Chemical Supplier No. 1487810 MG
A purine antimetabolite and an ADA inhibitor (Ki = 0.09 nM); reduces 2'-deoxyadenosine-induced growth inhibition of MOLT-4 lymphoblast cells (Ki = 5.6 µM); inhibits erythrocytes and ascites tumor cell ADA in an L1210 murine lymphocytic leukemia model at 0.2 mg/kg; inhibits tumor growth and increases survival rate in a CT26/NY-ESO-1 mouse xenograft model at 10 UG/mouse