Learn More
Cayman Chemical Pentostatin, 53910-25-1, 1 mg
SDP
Supplier:  Cayman Chemical 148781
Encompass_Preferred
A purine nucleoside analog that irreversibly inhibits adenosine deaminase (Ki = 0.9 pM) and thus interrupts DNA synthesis in dividing cells; displays strong efficacy in the clinical treatment of hairy cell leukemia as well as relapsed chronic lymphocytic leukemia
Catalog No. NC1577131