Learn More
FUJIFILM Biosciences Pentoxazone Standard
Catalog No. 5000450320
Encompass_Preferred
Pentoxazone Standard is a high-quality reference material designed for accurate quantification and analysis of pesticide residues With 95% purity it ensures consistent results in analytical methods like HPLC and qNMR Stable under proper storage conditions and easy to dissolve in acetone and ethanol
Catalog No. 50-004-50320 Supplier FUJIFILM Biosciences Supplier No. 16919691
Pentoxazone Standard is a high-quality reference material designed for accurate quantification and analysis of pesticide residues With 95% purity it ensures consistent results in analytical methods like HPLC and qNMR Stable under proper storage conditions and easy to dissolve in acetone and ethanol