Learn More
Medchemexpress LLC Pentoxifylline-d5 | 1185995-18-9 | 99.9% | 283.34 | C13H13D5N4O3 | 10 MG
SDP
Catalog No. 5000298113
Encompass_Preferred
Pentoxifylline-d5 is the deuterated analogue of pentoxifylline supplied for analytical and research use as an internal standard and tracer. It is intended for quantitative bioanalytical and pharmacokinetic studies and is compatible with NMR, GC-MS, and LC-MS methods.
  • Deuterated internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS assays.
  • High isotopic and chemical purity for accurate quantitation.
  • Provided in milligram-scale quantities for analytical workflows.
  • Useful in pharmacokinetic and tracer studies.
Catalog No. 50-002-98113 Supplier Medchemexpress LLC Supplier No. HYB0715S210MG
Pentoxifylline-d5 is the deuterated analogue of pentoxifylline supplied for analytical and research use as an internal standard and tracer. It is intended for quantitative bioanalytical and pharmacokinetic studies and is compatible with NMR, GC-MS, and LC-MS methods.
  • Deuterated internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS assays.
  • High isotopic and chemical purity for accurate quantitation.
  • Provided in milligram-scale quantities for analytical workflows.
  • Useful in pharmacokinetic and tracer studies.