Learn More
Medchemexpress LLC Pentoxifylline-d5 | 1185995-18-9 | 99.9% | 50 MG
SDP
Catalog No. 5000406754
Encompass_Preferred
Pentoxifylline-d5 is a deuterium-labeled version of Pentoxifylline. The unlabeled compound, Pentoxifylline (BL-191), acts as a haemorheological agent and is an orally active, non-selective phosphodiesterase (PDE) inhibitor. It demonstrates immune modulation, anti-inflammatory, hemorheological, anti-fibrinolytic, and anti-proliferation effects. This compound can be utilized in research concerning peripheral vascular disease, cerebrovascular disease, and other conditions involving impaired regional microcirculation.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development.
  • Deuteration can influence the pharmacokinetic and metabolic profiles of compounds.
Catalog No. 50-004-06754 Supplier Medchemexpress LLC Supplier No. HYB0715S250MG
Pentoxifylline-d5 is a deuterium-labeled version of Pentoxifylline. The unlabeled compound, Pentoxifylline (BL-191), acts as a haemorheological agent and is an orally active, non-selective phosphodiesterase (PDE) inhibitor. It demonstrates immune modulation, anti-inflammatory, hemorheological, anti-fibrinolytic, and anti-proliferation effects. This compound can be utilized in research concerning peripheral vascular disease, cerebrovascular disease, and other conditions involving impaired regional microcirculation.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development.
  • Deuteration can influence the pharmacokinetic and metabolic profiles of compounds.