Learn More
Medchemexpress LLC Pentoxifylline-d6 | 1185878-98-1 | 99.0% | 1 MG
SDP
Catalog No. 5000409379
Encompass_Preferred
Pentoxifylline-d6 is the deuterium-labeled Pentoxifylline. Pentoxifylline is a haemorheological agent and an orally active non-selective phosphodiesterase (PDE) inhibitor. It exhibits immune modulation, anti-inflammatory, hemorheological, anti-fibrinolytic, and anti-proliferation effects. Pentoxifylline is applicable for research in peripheral vascular disease, cerebrovascular disease, and other conditions involving defective regional microcirculation.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules serve as tracers for quantitation during drug development.
  • Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-09379 Supplier Medchemexpress LLC Supplier No. HYB0715S1MG
Pentoxifylline-d6 is the deuterium-labeled Pentoxifylline. Pentoxifylline is a haemorheological agent and an orally active non-selective phosphodiesterase (PDE) inhibitor. It exhibits immune modulation, anti-inflammatory, hemorheological, anti-fibrinolytic, and anti-proliferation effects. Pentoxifylline is applicable for research in peripheral vascular disease, cerebrovascular disease, and other conditions involving defective regional microcirculation.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules serve as tracers for quantitation during drug development.
  • Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.