Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pentoxifylline-d6 | 1185878-98-1 | 99.0% | 284.34 g/mol | C13H12D6N4O3 | 10 MG
SDP

Catalog No. 5000296728
Encompass_Preferred

Pentoxifylline-d6 is the deuterium-labeled analog of pentoxifylline provided as a research standard for analytical and tracer studies. It is intended for use as an internal standard in mass spectrometry, quantitative analysis, and pharmacokinetic or metabolic research; for laboratory research use only.

  • Deuterium-labeled (d6) internal standard for mass spectrometry.
  • High purity: 99.0%.
  • Molecular formula C13H12D6N4O3; molecular weight 284.34 g/mol.
  • CAS number 1185878-98-1 for unambiguous identification.
  • Appearance: white to off-white solid suitable for analytical handling.
  • Available as a 10 mg research quantity.

Catalog No. 50-002-96728 Supplier Medchemexpress LLC Supplier No. HYB0715S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pentoxifylline-d6 is the deuterium-labeled analog of pentoxifylline provided as a research standard for analytical and tracer studies. It is intended for use as an internal standard in mass spectrometry, quantitative analysis, and pharmacokinetic or metabolic research; for laboratory research use only.

  • Deuterium-labeled (d6) internal standard for mass spectrometry.
  • High purity: 99.0%.
  • Molecular formula C13H12D6N4O3; molecular weight 284.34 g/mol.
  • CAS number 1185878-98-1 for unambiguous identification.
  • Appearance: white to off-white solid suitable for analytical handling.
  • Available as a 10 mg research quantity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.