Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentoxifylline 98.0+%, TCI America™
SDP

Catalog No. P205025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P205025G 25 g
P20505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P205025G Supplier TCI America Supplier No. P205025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5-6-6493
Molecular Formula C13H18N4O3
Molecular Weight (g/mol) 278.31
MDL Number MFCD00063379
InChI Key BYPFEZZEUUWMEJ-UHFFFAOYSA-N
Synonym pentoxifylline, trental, oxpentifylline, pentoxifyllin, torental, pentoxyphylline, pentoxil, dimethyloxohexylxanthine, pentoxiphyllium, vazofirin
PubChem CID 4740
ChEBI CHEBI:7986
IUPAC Name 3,7-dimethyl-1-(5-oxohexyl)purine-2,6-dione
SMILES CC(=O)CCCCN1C(=O)C2=C(N=CN2C)N(C1=O)C

Specifications

Melting Point 105°C
Color White
Quantity 25 g
Formula Weight 278.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pentoxifylline
RTECSNumber XH2475000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.