Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentoxifylline, Thermo Scientific™

Catalog No. AC328430010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC328430010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC328430010 Supplier Thermo Scientific Chemicals Supplier No. 328430010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5-6-6493
Molecular Formula C13H18N4O3
Molecular Weight (g/mol) 278.31
InChI Key BYPFEZZEUUWMEJ-UHFFFAOYSA-N
Synonym pentoxifylline, trental, oxpentifylline, pentoxifyllin, torental, pentoxyphylline, pentoxil, dimethyloxohexylxanthine, pentoxiphyllium, vazofirin
PubChem CID 4740
ChEBI CHEBI:7986
IUPAC Name 3,7-dimethyl-1-(5-oxohexyl)purine-2,6-dione
SMILES CC(=O)CCCCN1C(=O)C2=C(N=CN2C)N(C1=O)C

Specifications

Color White
Infrared Spectrum Authentic
Quantity 1 g
Merck Index 15, 7249
Formula Weight 278.3
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material Pentoxifylline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.