Learn More
Medchemexpress LLC Pentoxyverine | 77-23-6 | MFCD00209899 | 98.9% | C20H31NO3 | 10MG
SDP
Catalog No. 5000258434
Encompass_Preferred
Pentoxyverine is an antitussive research reagent (carbetapentane) that acts as a sigma-1 receptor agonist and muscarinic antagonist. Supplied as a high-purity solid for laboratory use, it is used in pharmacology studies of cough reflex modulation, anticonvulsant effects, and airway smooth muscle activity.
  • High purity for research applications.
  • Active at sigma-1 and muscarinic receptors.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Available as small-mass solid and solution formats.
  • Supplied with a datasheet documenting identity and properties.
Catalog No. 50-002-58434 Supplier Medchemexpress LLC Supplier No. HY13400410MG
Pentoxyverine is an antitussive research reagent (carbetapentane) that acts as a sigma-1 receptor agonist and muscarinic antagonist. Supplied as a high-purity solid for laboratory use, it is used in pharmacology studies of cough reflex modulation, anticonvulsant effects, and airway smooth muscle activity.
  • High purity for research applications.
  • Active at sigma-1 and muscarinic receptors.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Available as small-mass solid and solution formats.
  • Supplied with a datasheet documenting identity and properties.