Learn More
Selleck Chemical LLC Pentoxyverine Citrate S4143-100mg
SDP
Catalog No. 503111195
Encompass_Preferred
Pentoxyverine Citrate (Carbetapentane) is an antitussive (cough suppressant) commonly used for cough associated with illnesses like common cold
Catalog No. 50-311-1195 Supplier Selleck Chemical LLC Supplier No. S4143100MG
Pentoxyverine Citrate (Carbetapentane) is an antitussive (cough suppressant) commonly used for cough associated with illnesses like common cold