Learn More
Medchemexpress LLC Pentoxyverine (citrate) | 23142-01-0 | 99.6% | 1 ML
SDP
Catalog No. 5000413975
Encompass_Preferred
Pentoxyverine citrate (Carbetapentane) is an orally active, blood-brain barrier permeable sigma-1 receptor agonist and muscarinic antagonist. It acts as a potent antitussive, anticonvulsant, and spasmolytic agent.
  • Orally active and blood-brain barrier permeable
  • Sigma-1 receptor agonist
  • Muscarinic antagonist
  • Potent antitussive, anticonvulsant, and spasmolytic agent
  • Inhibits bronchial interceptors
  • Weakens cough reflex
  • Relaxes bronchial smooth muscle
  • Reduces airway resistance
  • For research use only
Catalog No. 50-004-13975 Supplier Medchemexpress LLC Supplier No. HYB105510MM/1ML
Add to Cart
Add to Cart
Pentoxyverine citrate (Carbetapentane) is an orally active, blood-brain barrier permeable sigma-1 receptor agonist and muscarinic antagonist. It acts as a potent antitussive, anticonvulsant, and spasmolytic agent.
  • Orally active and blood-brain barrier permeable
  • Sigma-1 receptor agonist
  • Muscarinic antagonist
  • Potent antitussive, anticonvulsant, and spasmolytic agent
  • Inhibits bronchial interceptors
  • Weakens cough reflex
  • Relaxes bronchial smooth muscle
  • Reduces airway resistance
  • For research use only