Learn More
Description
- 55 assays on board; ISe model includes ISE module (Na+, K+, Cl- )
- Up to 420 tests/hr. throughput (with ISE module)
- Load up to 60 samples (6 racks containing 10 samples each)
- Touch-sceen integrated work and validation stations
- Uses Ready-to-use reagents; only one calibrator and 2 controls required for general chemistries
- Interfaces to lab information systems for automatic request and result transfer with permanent backup; stores up to 100,000 test results
Specifications
Specifications
| Type | Clinical Chemistry Analyzer |
| Sample Type | Whole Blood, Plasma, Serum, Urine, CSF, Homogeneous Fluids |
| Sample Dilution | 1/2 to 1/22,500 |
| Throughput | 420 Tests/hr. |
| Distilled Water Consumption | 10 L tank de-ionized/distilled water |
| Certifications/Compliance | NGSP |
| Quantity | 420 Tests |
| Height (English) | 25 in. |
| Height (Metric) | 63.5 cm |
| Width (English) | 40 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.