Learn More
Cayman Chemical Penetratin (trifluoroacetate salt), Purity: ≥95%, 5 mg
SDP
Catalog No. 502915045
Encompass_Preferred
A cell-penetrating peptide; has been used in the surface functionalization of liposomes for drug delivery across the BBB
Catalog No. 50-291-5045 Supplier Cayman Chemical Supplier No. 368785 MG
A cell-penetrating peptide; has been used in the surface functionalization of liposomes for drug delivery across the BBB