Learn More
Alpco Ltd Pentraxin 3 (PTX3) / TSG-14 ELISA
SDP
Supplier:  Alpco Ltd 48PTXHUE01
Encompass_Preferred
The Human Pentraxin3 / TSG-14 ELISA is a solid-phase sandwich immunoassay (ELISA) capable of measuring human PTX3 levels in plasma and serum free cell culture supernatant. For Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. NC2329191