Learn More
Medchemexpress LLC Pentyl 4-hydroxybenzoate | 6521-29-5 | MFCD00016479 | 98.2% | 208.25 g/mol | C12H16O3 | 25 MG
SDP
Catalog No. 5000282432
Encompass_Preferred
Pentyl 4-hydroxybenzoate (Amylparaben) is the pentyl ester of 4-hydroxybenzoic acid used as an antimicrobial preservative and research reagent. It is supplied in small pack sizes for analytical and laboratory work and is accompanied by manufacturer documentation for quality and safe handling.
  • High purity (98.18%) suitable for analytical applications.
  • Acts as a preservative and antimicrobial agent in formulations and studies.
  • Provided with datasheet, certificate of analysis, and safety data sheet for traceability.
  • Available in small pack sizes for laboratory use (25 MG to 500 MG).
  • Chemical formula C12H16O3 and molecular weight 208.25 g/mol for accurate formulation.
Catalog No. 50-002-82432 Supplier Medchemexpress LLC Supplier No. HYW26744425MG
Pentyl 4-hydroxybenzoate (Amylparaben) is the pentyl ester of 4-hydroxybenzoic acid used as an antimicrobial preservative and research reagent. It is supplied in small pack sizes for analytical and laboratory work and is accompanied by manufacturer documentation for quality and safe handling.
  • High purity (98.18%) suitable for analytical applications.
  • Acts as a preservative and antimicrobial agent in formulations and studies.
  • Provided with datasheet, certificate of analysis, and safety data sheet for traceability.
  • Available in small pack sizes for laboratory use (25 MG to 500 MG).
  • Chemical formula C12H16O3 and molecular weight 208.25 g/mol for accurate formulation.