Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pentyl 4-hydroxybenzoate | 6521-29-5 | MFCD00016479 | 98.2% | 208.25 g/mol | C12H16O3 | 50 MG
SDP

Catalog No. 5000282417
Encompass_Preferred

Pentyl 4-hydroxybenzoate (Amylparaben) is a paraben-class preservative supplied as a white to off-white solid for research and analytical applications. It is commonly used in antimicrobial assays, formulation testing, and as an analytical reference standard, and is provided with specified purity and storage conditions for laboratory use.

  • High purity: 98.2%.
  • Physical form: white to off-white solid.
  • Molecular weight: 208.25 g/mol.
  • Chemical formula: C12H16O3.
  • Storage recommendations: 4°C under nitrogen; in solvent store at -80°C for long-term.
  • Suitable for antimicrobial and formulation studies and analytical reference use.

Catalog No. 50-002-82417 Supplier Medchemexpress LLC Supplier No. HYW26744450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pentyl 4-hydroxybenzoate (Amylparaben) is a paraben-class preservative supplied as a white to off-white solid for research and analytical applications. It is commonly used in antimicrobial assays, formulation testing, and as an analytical reference standard, and is provided with specified purity and storage conditions for laboratory use.

  • High purity: 98.2%.
  • Physical form: white to off-white solid.
  • Molecular weight: 208.25 g/mol.
  • Chemical formula: C12H16O3.
  • Storage recommendations: 4°C under nitrogen; in solvent store at -80°C for long-term.
  • Suitable for antimicrobial and formulation studies and analytical reference use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.