Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pentyl Acetate

Catalog No. 460221LF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-022-1LF 1 L
46-022-250MLF 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 46-022-1LF Supplier Sigma Aldrich Supplier No. 460221LF
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Pentyl Acetate

Specifications

CAS 628-63-7
Melting Point -100°C
Density 0.876 g/mL at 25°C
Boiling Point 142°C to 149°C
Molecular Formula C7H14O2
Refractive Index n20/D 1.402; n20/D 1.403
Linear Formula CH3COO(CH2)4CH3
MDL Number MFCD00009500
Quantity 1 L
Synonym Amyl acetate
Molecular Weight (g/mol) 130.18 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.