Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentyl acetate, Analytical Standard, ≥97.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019715 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9715 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9715 Supplier MilliporeSigma Supelco Supplier No. 669621ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 628-63-7
Vapor Pressure 4 mmHg (20°C)
Vapor Density 4.5 (vs. air)
Density 0.876 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3COO(CH2)4 CH3
MDL Number MFCD00009500
Melting Point -100°C
Boiling Point 142°C to 149°C
Synonym Amyl acetate
Quantity 1 mL
Formula Weight 130.18
Percent Purity ≥97.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.