Learn More
Medchemexpress LLC Pentyl octanoate | 638-25-5 | 98.0% | 214.34 g/mol | C13H26O2 | 100 G
SDP
Catalog No. 5000282643
Encompass_Preferred
Pentyl octanoate (Amyl n-Octanoate) is an organic ester used as a biochemical reagent and solvent in laboratory research. It has the molecular formula C13H26O2, CAS 638-25-5, and a molecular weight of 214.34 g/mol. The compound is typically supplied as a neat liquid in multiple pack sizes for chemical and life-science applications; check the supplier certificate of analysis for an authoritative purity value.
  • Common applications include solvent, reagent, and fragrance intermediate.
  • Available in multiple pack sizes, including 100 g.
  • Chemical identity: C13H26O2; CAS 638-25-5.
  • Physical state is a liquid suitable for laboratory handling.
  • Store at room temperature in a cool, dry place; in solvent, long-term storage at -20 °C to -80 °C is reported.
  • Confirm purity and handling details via the supplier certificate of analysis or safety data sheet.
Catalog No. 50-002-82643 Supplier Medchemexpress LLC Supplier No. HYW127548100G
Pentyl octanoate (Amyl n-Octanoate) is an organic ester used as a biochemical reagent and solvent in laboratory research. It has the molecular formula C13H26O2, CAS 638-25-5, and a molecular weight of 214.34 g/mol. The compound is typically supplied as a neat liquid in multiple pack sizes for chemical and life-science applications; check the supplier certificate of analysis for an authoritative purity value.
  • Common applications include solvent, reagent, and fragrance intermediate.
  • Available in multiple pack sizes, including 100 g.
  • Chemical identity: C13H26O2; CAS 638-25-5.
  • Physical state is a liquid suitable for laboratory handling.
  • Store at room temperature in a cool, dry place; in solvent, long-term storage at -20 °C to -80 °C is reported.
  • Confirm purity and handling details via the supplier certificate of analysis or safety data sheet.