Learn More
Selleck Chemical LLC Pentylenetetrazol 100mg 54-95-5 Pentylenetetrazole, Metrazole, pentetrazol, pentamethylenetetrazol, Corazol, Cardiazol, PTZ
SDP
Catalog No. 501364026
Encompass_Preferred
Pentylenetetrazol (Pentylenetetrazole, Metrazole, pentetrazol, pentamethylenetetrazol, Corazol, Cardiazol, PTZ) displays activity as a central nervous system and respiratory stimulant. It is considered a non-competitive GABA(A) receptor antagonist. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4026 Supplier Selleck Chemical LLC Supplier No. S4587100MG
Pentylenetetrazol (Pentylenetetrazole, Metrazole, pentetrazol, pentamethylenetetrazol, Corazol, Cardiazol, PTZ) displays activity as a central nervous system and respiratory stimulant. It is considered a non-competitive GABA(A) receptor antagonist. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.