Learn More
Medchemexpress LLC Pentynoic acid STP ester sodium | 1807530-14-8 | MFCD26793735 | 99.5% | 348.20 g·mol⁻¹ | C11H5F4NaO5S | 5 MG
SDP
Catalog No. 5000223386
Encompass_Preferred
Pentynoic acid STP ester sodium is a water-soluble click-chemistry reagent that contains a terminal propargyl (alkyne) group and an STP (sulfotetrafluorophenyl) activated ester. It is used to introduce alkyne handles onto biomolecules via amine-directed conjugation, enabling subsequent bioorthogonal click reactions.
  • Water-soluble alkyne-activated ester for bioconjugation.
  • Enables installation of terminal alkyne handles on proteins and peptides.
  • STP activated ester reacts efficiently with primary amines.
  • High reported purity suitable for sensitive biomolecular applications.
  • Stable when stored sealed and protected from moisture; compatible with low-temperature storage in solution.
Catalog No. 50-002-23386 Supplier Medchemexpress LLC Supplier No. HY1382465MG
Pentynoic acid STP ester sodium is a water-soluble click-chemistry reagent that contains a terminal propargyl (alkyne) group and an STP (sulfotetrafluorophenyl) activated ester. It is used to introduce alkyne handles onto biomolecules via amine-directed conjugation, enabling subsequent bioorthogonal click reactions.
  • Water-soluble alkyne-activated ester for bioconjugation.
  • Enables installation of terminal alkyne handles on proteins and peptides.
  • STP activated ester reacts efficiently with primary amines.
  • High reported purity suitable for sensitive biomolecular applications.
  • Stable when stored sealed and protected from moisture; compatible with low-temperature storage in solution.