Learn More
Medchemexpress LLC HY-112730 10mg Medchemexpress, PEO-IAA CAS:6266-66-6 Purity:>98%
SDP
Catalog No. 501969160
Encompass_Preferred
Medchemexpress, HY-112730 10mg PEO-IAA CAS:6266-66-6 PEO-IAA is an indole-3-acetic acid (IAA) antagonist. PEO-IAA is an auxin antagonist that binds to transport inhibitor response 1/auxin signaling F-box proteins (TIR1/AFBs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9160 Supplier Medchemexpress LLC Supplier No. HY11273010MG
Medchemexpress, HY-112730 10mg PEO-IAA CAS:6266-66-6 PEO-IAA is an indole-3-acetic acid (IAA) antagonist. PEO-IAA is an auxin antagonist that binds to transport inhibitor response 1/auxin signaling F-box proteins (TIR1/AFBs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.