Learn More
Aviva Systems Biology Corp TWNK Antibody - middle region (ARP36483_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36483_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PEO1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PEO1. Alternate names: C10orf2, FLJ21832, PEO, PEOA3, SANDO, TWINL, PEO1, SCA8, ATXN8, IOSCA, MTDPS7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53049