Learn More
Cayman Chemical (PEO)3-monoamine, Purity: ≥98%, CAS: 6338-55-2, 250 mg
SDP
Catalog No. 502988086
Encompass_Preferred
A blocking reagent; prevents nonspecific adsorption of analytes in applications using covalent protein coupling; reduces the adsorption of IgG when coated on carboxylic-terminated surfaces
Catalog No. 50-298-8086 Supplier Cayman Chemical Supplier No. 30426250 MG
A blocking reagent; prevents nonspecific adsorption of analytes in applications using covalent protein coupling; reduces the adsorption of IgG when coated on carboxylic-terminated surfaces