Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Peonidin-3-O-arabinoside chloride | 524943-91-7 | 98.5% | 468.84 | 10 MG
SDP

Catalog No. 5000438038
Encompass_Preferred

Peonidin-3-O-arabinoside chloride is an anthocyanin that can be isolated from Vaccinium myrtillus L. This product is for research use only.

  • For research use only
  • Store at 4°C, sealed, away from moisture
  • Stable in solvent for 6 months at -80°C or 1 month at -20°C
  • Appearance: Solid, brown to black
  • Molecular formula: C21H21ClO10
  • Structure classification: Phenols, polyphenols

Catalog No. 50-004-38038 Supplier Medchemexpress LLC Supplier No. HY12641310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Peonidin-3-O-arabinoside chloride is an anthocyanin that can be isolated from Vaccinium myrtillus L. This product is for research use only.

  • For research use only
  • Store at 4°C, sealed, away from moisture
  • Stable in solvent for 6 months at -80°C or 1 month at -20°C
  • Appearance: Solid, brown to black
  • Molecular formula: C21H21ClO10
  • Structure classification: Phenols, polyphenols

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.