Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Peonidin-3-O-arabinoside chloride | 524943-91-7 | 98.5% | 468.84 | 25 MG
SDP

Catalog No. 5000439648
Encompass_Preferred

Peonidin-3-O-arabinoside chloride is an anthocyanin that can be isolated from *Vaccinium myrtillus L.*. This chemical compound is available as a solid with high purity, suitable for various research applications.

  • High purity: 98.5%
  • Solid appearance
  • Brown to black color
  • Recommended storage at 4°C, sealed, away from moisture
  • Extended storage in solvent at -80°C (6 months) or -20°C (1 month)

Catalog No. 50-004-39648 Supplier Medchemexpress LLC Supplier No. 25MGHY126413
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Peonidin-3-O-arabinoside chloride is an anthocyanin that can be isolated from *Vaccinium myrtillus L.*. This chemical compound is available as a solid with high purity, suitable for various research applications.

  • High purity: 98.5%
  • Solid appearance
  • Brown to black color
  • Recommended storage at 4°C, sealed, away from moisture
  • Extended storage in solvent at -80°C (6 months) or -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.