Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Peonidin-3-O-arabinoside chloride | 524943-91-7 | MFCD03427688 | 98.5% | 468.84 g/mol | C21H21ClO10 | 5 MG
SDP

Catalog No. 5000264840
Encompass_Preferred

Peonidin-3-O-arabinoside chloride is an anthocyanin chloride salt isolated from Vaccinium myrtillus L. It is provided as a high-purity solid for biochemical and pharmacological research, suitable for analytical assays and small-scale experimental use.

  • Purity 98.5%.
  • Chemical formula C21H21ClO10.
  • Molecular weight 468.84 g/mol.
  • Physical form solid.
  • Supplied in milligram-scale quantities for research use.
  • Natural source documented from Vaccinium myrtillus L.

Catalog No. 50-002-64840 Supplier Medchemexpress LLC Supplier No. HY1264135MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Peonidin-3-O-arabinoside chloride is an anthocyanin chloride salt isolated from Vaccinium myrtillus L. It is provided as a high-purity solid for biochemical and pharmacological research, suitable for analytical assays and small-scale experimental use.

  • Purity 98.5%.
  • Chemical formula C21H21ClO10.
  • Molecular weight 468.84 g/mol.
  • Physical form solid.
  • Supplied in milligram-scale quantities for research use.
  • Natural source documented from Vaccinium myrtillus L.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.