Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Peonidin-3-O-galactoside (chloride) | 28148-89-2 | 97.1% | 498.86 g/mol | C22H23ClO11 | 5 MG
SDP

Catalog No. 5000264837
Encompass_Preferred

Peonidin-3-O-galactoside chloride is an anthocyanin glycoside with reported antioxidant properties. It is supplied as a brown to black solid for research and analytical use and is intended as a reference standard in biochemical and natural product studies.

  • Anthocyanin glycoside with antioxidant activity.
  • CAS number 28148-89-2.
  • Molecular formula C22H23ClO11.
  • Molecular weight 498.86 g/mol.
  • Assay purity 97.1%.
  • Brown to black solid appearance.
  • Store sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-64837 Supplier Medchemexpress LLC Supplier No. HY1264115MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Peonidin-3-O-galactoside chloride is an anthocyanin glycoside with reported antioxidant properties. It is supplied as a brown to black solid for research and analytical use and is intended as a reference standard in biochemical and natural product studies.

  • Anthocyanin glycoside with antioxidant activity.
  • CAS number 28148-89-2.
  • Molecular formula C22H23ClO11.
  • Molecular weight 498.86 g/mol.
  • Assay purity 97.1%.
  • Brown to black solid appearance.
  • Store sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.