Learn More
Medchemexpress LLC Peonidin chloride | 134-01-0 | 5 MG
SDP
Catalog No. 5000420974
Encompass_Preferred
Peonidin chloride is an anthocyanin compound found in plants such as grapes and purple-fleshed sweet potatoes. It does not exert a stable inhibitory effect on LPS-mediated inflammatory gene expression alone, but its acylated form exhibits antimutagenic activity. This product is for research use only.
  • Anthocyanin compound
  • Found in plants such as grapes and purple-fleshed sweet potatoes
  • Acylated form exhibits antimutagenic activity
  • Reduces LPS-induced inflammatory gene expression and upregulates HSL expression in primary human adipocytes when used in combination with malvidin
  • For research use only
Catalog No. 50-004-20974 Supplier Medchemexpress LLC Supplier No. HYN24595MG
Peonidin chloride is an anthocyanin compound found in plants such as grapes and purple-fleshed sweet potatoes. It does not exert a stable inhibitory effect on LPS-mediated inflammatory gene expression alone, but its acylated form exhibits antimutagenic activity. This product is for research use only.
  • Anthocyanin compound
  • Found in plants such as grapes and purple-fleshed sweet potatoes
  • Acylated form exhibits antimutagenic activity
  • Reduces LPS-induced inflammatory gene expression and upregulates HSL expression in primary human adipocytes when used in combination with malvidin
  • For research use only