Learn More
CPC Scientific H-Lys-Glu-Thr-Trp-Trp-Glu-Thr-Trp-Trp-Thr-Glu-Trp-Ser-Gln-Pro-Lys-Lys-Lys-Arg-Lys-Val-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific TAT001B
Encompass
Pep-1 is a cell-penetrating peptide. ONE-LETTER SEQUENCE: KETWWETWWTEWSQPKKKRKVMOLECULAR FORMULA: C136H195N35O33MOLECULAR WEIGHT:2848.22STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [395069-86-0], SYNONYMS: Pep-1 (uncapped)RESEARCH AREA: AntimicrobialREFERENCES: M. C. Morris, et al., Nature Pub. Group, Vol 19, pg 1173 (2001)
Catalog No. 50-211-0538