Learn More
Medchemexpress LLC HY-P2276 5mg , Pep2-8 CAS:1541011-97-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP22765MG
Encompass_Preferred
HY-P2276 5mg Pep2-8 CAS: 1541011-97-5 Pep2-8 is a PCSK9 inhibitor with a binding KD of 0.7 μM and an IC50 of 1.4 μM.
Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1579