Learn More
Medchemexpress LLC Pep42 TFA | 99.6% | 1335.59 | 1 MG
SDP
Catalog No. 5000365838
Encompass_Preferred
Pep42 TFA is a cyclic peptide ligand of GPR78. It internalizes selectively into highly metastatic human melanoma cells through a receptor-mediated process. This product is for research use only and not sold to patients.
  • Cyclic peptide ligand of GPR78
  • Selectively internalizes into highly metastatic human melanoma cells
Catalog No. 50-003-65838 Supplier Medchemexpress LLC Supplier No. 1MGHYP10374A
Pep42 TFA is a cyclic peptide ligand of GPR78. It internalizes selectively into highly metastatic human melanoma cells through a receptor-mediated process. This product is for research use only and not sold to patients.
  • Cyclic peptide ligand of GPR78
  • Selectively internalizes into highly metastatic human melanoma cells