Learn More
Aviva Systems Biology Corp Pepd Antibody - middle region (ARP56087_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56087_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pepd. It was validated on Western Blot. . Alternate names: Pep-4, Pep4ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-115