Learn More
Medchemexpress LLC Pepinh-TRIF TFA | 99.4% | 3875.54 | 10 MG
SDP
Catalog No. 5000422941
Encompass_Preferred
Pepinh-TRIF (TFA) is a 30 amino acid peptide designed to inhibit TIR-domain-containing adapter-inducing interferon-β (TRIF) signaling by disrupting the interaction between TLR and TRIF. This product is intended for research use only.
  • Inhibits TIR-domain-containing adapter-inducing interferon-β (TRIF) signaling
  • Disrupts interaction between TLR and TRIF
  • Blocks expression and production of IL-33 stimulated by polyI:C or flagellin
  • Suppresses stimulated IκB-α phosphorylation and degradation in HCECs
  • Suppresses NF-κB activation with p65 protein nuclear translocation
Catalog No. 50-004-22941 Supplier Medchemexpress LLC Supplier No. HYP256510MG
Pepinh-TRIF (TFA) is a 30 amino acid peptide designed to inhibit TIR-domain-containing adapter-inducing interferon-β (TRIF) signaling by disrupting the interaction between TLR and TRIF. This product is intended for research use only.
  • Inhibits TIR-domain-containing adapter-inducing interferon-β (TRIF) signaling
  • Disrupts interaction between TLR and TRIF
  • Blocks expression and production of IL-33 stimulated by polyI:C or flagellin
  • Suppresses stimulated IκB-α phosphorylation and degradation in HCECs
  • Suppresses NF-κB activation with p65 protein nuclear translocation