Learn More
Gibco™ PeproGMP™ Human Activin A Recombinant Protein, PeproTech®
Description
PeproTech Recombinant Human Activin A is a 26.0 kDa disulfide-linked homodimer of two beta-A chains, each containing 116 amino acid residues. Reconstitute using sterile water (i.e., WFI) to a concentration of 0.1-1.0 mg/ml. Additional product information, specifications, handling and reconstitution information are provided on the Certificate of Analysis. All PeproGMP™ products are manufactured using animal-derived component free materials. Product Description/Intended Use: PeproTech GMP recombinant proteins are manufactured for use as ancillary materials by applying applicable principles of GMP and quality control requirements from USP (United States Pharmacopeia) Chapter 1043 Ancillary Materials for Cell, Gene, and Tissue-Engineered Products.
Specifications
Specifications
| Accession Number | P08476 |
| For Use With (Application) | Control, Western Blot Control, Bioactivity |
| Format | Lyophilized |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 3624 |
| Molecular Weight (g/mol) | 26 kDa |
| Name | PeproGMP™ Human Activin A |
| Purification Method | Purified |
| Quantity | 100 μg |
| Storage Requirements | -20°C or -80°C if preferred |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.