Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ PeproGMP™ Human IL-7 Recombinant Protein, PeproTech®

Catalog No. 502558481
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-255-8481 100 μg
50-003-31917 500 μg
01-672-622 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-255-8481 Supplier Gibco™ Supplier No. GMP20007100UG
Only null left
Add to Cart
Add to Cart

Recombinant Protein

PeproTech Recombinant Human IL-7 is a 17.4 kDa protein containing 152 amino acid residues. PeproGMP IL-7 can be used with PeproGMP IL-15 in CTS NK-Xpander medium for natural killer (NK) cell differentiation from pluripotent stem cells. Reconstitute using sterile water (i.e., WFI) to a concentration of 0.1-1.0 mg/ml. Additional product information, specifications, handling and reconstitution information are provided on the Certificate of Analysis. All PeproGMP™ products are manufactured using animal-derived component free materials. Product Description/Intended Use: PeproTech GMP recombinant proteins are manufactured for use as ancillary materials by applying applicable principles of GMP and quality control requirements from USP (United States Pharmacopeia) Chapter 1043 Ancillary Materials for Cell, Gene, and Tissue-Engineered Products.

Interleukin 7 (IL-7) is a hematopoietic cytokine that is constitutively produced by stromal cells from the bone marrow and thymus. IL-7 signals through the IL-7R to promote the differentiation of hematopoietic stem cells to lymphoid precursor cells, which give rise to T cells, B cells, and natural killer cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P13232, P16871
For Use With (Application) Control, Western Blot Control, Bioactivity
Format Lyophilized
Formulation Protein with no preservative
Gene ID (Entrez) 3574, 3575
Molecular Weight (g/mol) 17.4 kDa
Name PeproGMP™ Human IL-7
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C or -80°C if preferred
Regulatory Status GMP
Endotoxin Concentration ≤0.1 EU/μg
Gene Alias A630026I06Rik; H-IL-7; hlb368; IL7; Il-7; ILN; Interleukin; interleukin 7; Interleukin7; interleukin-7; M-IL-7
Common Name IL-7
Gene Symbol Il7, IL7r
Biological Activity Determined by the proliferation of mouse 2E8 cells. PeproGMP Recombinant Human IL-7 ha been tested against the WHO standard NIBSC 90/530.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence DCDIEGKDGK QYESVLMVSI DQLLDSMKEI GSNCLNNEFN FFKRHICDAN KEGMFLFRAA RKLRQFLKMN STGDFDLHLL KVSEGTTILL NCTGQVKGRK PAALGEAQPT KSLEENKSLK EQKKLNDLCF LKRLLQEIKT CWNKILMGTK EH
Content And Storage -20°Cor to +80°C if preferred
Activity Determined by the proliferation of mouse 2E8 cells. PeproGMP Recombinant Human IL-7 has been tested against the WHO standard NIBSC 90/530. Find the International Unit (IU) value <a href='https://www.thermofisher.com/us/en/home/life-science/cell-culture/cell-culture-learning-center/recombinant-protein-information/international-units.html'>here</a>.
Expression System E. coli
Form Lyophilized
Product Line PeproGMP
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.