Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ PeproGMP® Human SCF Recombinant Protein, PeproTech®

Catalog No. 01672631
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-672-631 50 μg
50-255-8474 1 mg
50-255-8475 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-672-631 Supplier Gibco™ Supplier No. GMP3000750UG
  • $898.00
    $853.65 / Each of 1

    Save $44.35

    5% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

The human SCF gene encodes for a 273 amino acid transmembrane protein, which contains a 25 amino acid N-terminal signal sequence, a 189 amino acid extracellular domain, a 23 amino acid transmembrane domain, and a 36 amino acid cytoplasmic domain. The secreted soluble form of SCF is generated by proteolytic processing of the membrane anchored precursor. Additional product information and specifications are provided on the Certificate of Analysis. All PeproGMP® products are manufactured using animal-derived component free materials. Product Description/Intended Use: PeproTech GMP recombinant proteins are manufactured for use as ancillary materials by applying applicable principles of GMP and quality control requirements from USP (United States Pharmacopeia) Chapter 1043 Ancillary Materials for Cell, Gene, and Tissue-Engineered Products. Product use statement: For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.

Stem cell factor (SCF) is the ligand of the c-Kit oncogene and is expressed by various structural and inflammatory cells in the airways. Binding of SCF by the c-Kit receptor leads to homodimerization of the receptor and the activation of signalling pathways such as PI-3, PLC-gamma, Jak/STAT, and MAP kinase pathways. SCF expression leads to the induction of mast cell survival and the expression and release of histamine, pro-inflammatory cytokines and chemokines. The inhibition of the SCF/c-Kit pathway leads to a decrease in histamine levels, mast cell and eosinophil infiltration, IL-4 production and airway hyperresponsiveness, suggesting this pathway may be a useful therapeutic target in inflammatory diseases such as asthma. At least two isoforms of SCF are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P21583
For Use With (Application) Control, Western Blot Control, Bioactivity
Format Lyophilized
Formulation Protein with no preservative
Gene ID (Entrez) 4254
Molecular Weight (g/mol) 18.4 kDa
Name PeproGMP™ Human SCF
Purification Method Purified
Quantity 50 μg
Storage Requirements -20°C or -80°C if preferred
Regulatory Status GMP
Endotoxin Concentration ≤0.1 EU/μg
Gene Alias 710-712; blz; c-Kit ligand; Clo; cloud gray; Con; contrasted; CSF; DCUA; DFNA69; DKFZp686F2250; familial progressive hyperpigmentation 2; FPH2; FPHH; Gb; grizzle-belly; hematopoietic growth factor KL; H-SCF; Kit ligand; Kitl; Kitlg; kit-ligand; KL-1; Mast cell growth factor; MGF; M-SCF; Processed kit ligand; SCF; SF; SHEP7; sKITLG; Sl; SLF; Soluble KIT ligand; Steel factor; steel factor/kit ligand; stem cell factor; stem cell factor KL-1; stem cell factor, CSF
Common Name SCF
Gene Symbol KITLG
Biological Activity Determined by the proliferation of TF-1 cells. PeproGMP Recombinant Human SCF has been tested against the WHO standard NIBSC 91/682.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MEGICRNRVT NNVKDVTKLV ANLPKDYMIT LKYVPGMDVL PSHCWISEMV VQLSDSLTDL LDKFSNISEG LSNYSIIDKL VNIVDDLVEC VKENSSKDLK KSFKSPEPRL FTPEEFFRIF NRSIDAFKDF VVASETSDCV VSSTLSPEKD SRVSVTKPFM LPPVA
Content And Storage -20°Cor to +80°C if preferred
Activity Determined by the proliferation of TF-1 cells. PeproGMP Recombinant Human SCF has been tested against the WHO standard NIBSC 91/682. Find the International Unit (IU) value <a href='https://www.thermofisher.com/us/en/home/life-science/cell-culture/cell-culture-learning-center/recombinant-protein-information/international-units.html'>here</a>.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.