Learn More
Bruker Daltonics PEPSEP C18 50CMX75UM,1.5UM
Catalog No. NC2258801
Encompass
PepSep C18 50cmx75μm,1.5μm
This item is not returnable. View return policy
Catalog No. NC2258801 Supplier Bruker Daltonics Supplier No. 1895846
PepSep C18 50cmx75μm,1.5μm