Learn More
Sigma Aldrich Fine Chemicals Biosciences Pepsin, 1g
Catalog No. NC1722130
Encompass
Pepsin is an aspartic endoproteinase isolated from pig gastric mucosa. It is mainly used for the unspecific hydrolysis of proteins and peptides in acidic media. It also provides a limited hydrolysis of native immunoglobulins to yield biologically active fragments.
Catalog No. NC1722130 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10108057001
Pepsin is an aspartic endoproteinase isolated from pig gastric mucosa. It is mainly used for the unspecific hydrolysis of proteins and peptides in acidic media. It also provides a limited hydrolysis of native immunoglobulins to yield biologically active fragments.