Learn More
Medchemexpress LLC Pepsin | 9001-75-6 | 250 MG
SDP
Catalog No. 5000426237
Encompass_Preferred
Pepsin is the major pig and human gastric protease and a minor pepsin-like gastric proteolytic enzyme. It contributes to the proteolysis of food proteins in the vertebrate stomach.
  • Optimal pH for activity is 1.0-3.0.
  • Activated by hydrochloric acid (HCl) and trifluoroacetic acid (TFA).
  • Inactivated by pH greater than 6.0.
  • To reconstitute, resuspend in double-distilled water (pH 5.5 or lower) to 1 mg/mL.
  • Reconstituted Pepsin can be stored at 4 °C for up to one month.
  • Cleavage specificity for Phenylalanine (Phe) and Leucine (Leu) is best at pH 1.0 and decreased above pH 2.0.
  • Irreversibly inactivates above pH 6.0.
Catalog No. 50-004-26237 Supplier Medchemexpress LLC Supplier No. 250MGHYP1635
Pepsin is the major pig and human gastric protease and a minor pepsin-like gastric proteolytic enzyme. It contributes to the proteolysis of food proteins in the vertebrate stomach.
  • Optimal pH for activity is 1.0-3.0.
  • Activated by hydrochloric acid (HCl) and trifluoroacetic acid (TFA).
  • Inactivated by pH greater than 6.0.
  • To reconstitute, resuspend in double-distilled water (pH 5.5 or lower) to 1 mg/mL.
  • Reconstituted Pepsin can be stored at 4 °C for up to one month.
  • Cleavage specificity for Phenylalanine (Phe) and Leucine (Leu) is best at pH 1.0 and decreased above pH 2.0.
  • Irreversibly inactivates above pH 6.0.