Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Pepsin, for biochemistry, powder

Catalog No. AC417071000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC417071000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC417071000 Supplier Thermo Scientific Chemicals Supplier No. 417071000

Pepsin from porcine gastric mucosa, CAS # 9001-75-6, is a peptidase that is commonly used in enzymatic digestion of proteins. | CAS: 9001-75-6

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Pepsin is an endopeptidase that digests proteins by hydrolyzing peptide bonds
  • It is the active form of pepsinogen, which is produced in the chief cells of the gastric mucosa

Applications

  • Pepsin is commonly used to produce F(ab’)2 fragments from antibodies
  • It has been used along with other enzymes to demonstrate the effects of fixation and enzymatic digestion in immunohistochemical assays, using paraffin-embedded tissue
  • It can be used in hydrolyzing dry cervical samples in mice
  • It has been used in the digestion of crude wheat gliadin
  • It has been used to stimulate in vitro gastric digestion of cooked cod, cocoa mass, and dietary fiber
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9001-75-6

Specifications

Quantity 100 g
Product Type Pepsin
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement:
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.