Learn More
MOLECULAR DEPOT LLC Pepsin Digested BSA - 50 mL
Catalog No. 5000013328
Encompass_Preferred
Pepsin Digested BSA consists of an enzyme-hydrolyzed version of BSA with various applications in biochemistry reagents formulation and more Pepsin-hydrolyzed BSA consists of a collection of small peptides that can minimize non-specific interactions For in vitro research use only
Catalog No. 50-000-13328 Supplier MOLECULAR DEPOT LLC Supplier No. P2010031
Pepsin Digested BSA consists of an enzyme-hydrolyzed version of BSA with various applications in biochemistry reagents formulation and more Pepsin-hydrolyzed BSA consists of a collection of small peptides that can minimize non-specific interactions For in vitro research use only