Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Pepsin From Porcine Stomach Mucosa

Catalog No. ICN10259890 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10259890 500 g
ICN10259850 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10259890 Supplier MP Biomedicals™ Supplier No. 0210259890
Add to Cart
Add to Cart

Pepsin cleaves peptides with an aromatic acid on either side of the peptide bond

Pepsin contains the “cathepsin” component which has milk curdling activity. It has a broad range of substrate activity and demonstrates an esterase acitivity.

  • CAS Number: 9001-75-6
  • EC Number: 232-629-3
  • MDL Number: MFCD00081840
  • Format: off white lyophilized powder
  • Specific activity: ≥1:10000

Used in preparation of Fab fragments from antibodies and to hydrolyze dry cervical samples in mice

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Molecular Biology, Proteomics
Molecular Weight (g/mol) ∽35000
Name Pepsin
pH Range 2.5 to 4.5
Quantity 500 g
Source Porcine
Storage Requirements 15° to 30°C
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.