Learn More
Mettler Toledo™ Pepsin-HCl Cleaning Solution
Catalog No. 01912587 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Size:
250 mL
Catalog No. Size
01-912-587 250 mL
1 options
Catalog No. 01-912-587 Supplier Mettler Toledo™ Supplier No. 51350100
Add to Cart
Add to Cart
Electrode cleaning solution
  • Pepsin-HCl Cleaning Solution
  • Keeps electrodes ready with the right cleaning, storage, and regeneration solutions
  • 250 mL
TRUSTED_SUSTAINABILITY
Specifications
Description Pepsin, HCl
Type Electrodes Cleaner
For Use With (Application) Cleaning Electrodes
Size 250 mL