Learn More
Medchemexpress LLC Pepsin (MS Grade) | 9001-75-6 | 250 MG
SDP
Catalog No. 5000431245
Encompass_Preferred
Pepsin (MS Grade) is a major pig and human gastric protease, functioning as a pepsin-like minor gastric proteolytic enzyme. It contributes to the proteolysis of food proteins in the vertebrate stomach. This product is suitable for:
  • Protein analysis in mass spectrometry and other applications
  • Structural protein research (HDX exchange) and antibody analysis
  • Digestion of tightly folded proteins in proteomics
  • Digestion of casein for vaccine production protein
  • Hydrolyzing hemoglobin to produce 1 μmol tyrosine per minute at 37°C
  • Storage at 4°C for 2 years
Catalog No. 50-004-31245 Supplier Medchemexpress LLC Supplier No. 250MGHYE70198
Pepsin (MS Grade) is a major pig and human gastric protease, functioning as a pepsin-like minor gastric proteolytic enzyme. It contributes to the proteolysis of food proteins in the vertebrate stomach. This product is suitable for:
  • Protein analysis in mass spectrometry and other applications
  • Structural protein research (HDX exchange) and antibody analysis
  • Digestion of tightly folded proteins in proteomics
  • Digestion of casein for vaccine production protein
  • Hydrolyzing hemoglobin to produce 1 μmol tyrosine per minute at 37°C
  • Storage at 4°C for 2 years