Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Pepsin, from Porcine

Catalog No. MP210259880 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210259880 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210259880 Supplier MP Biomedicals™ Supplier No. 0210259880
Add to Cart
Add to Cart

Pepsin, from Porcine≥1:10,000, Off-white Lyophilized Powder

  • Pepsin is a peptidase used to digest proteins. It is commonly used in the preparation of Fab fragments from antibodies. Pepsin, from porcine gastric mucosa, has been used to hydrolyze dry cervical samples in mice. Pepsin, an acid protease, contains a proteolytic enzyme. Pepsin contains the “cathepsin” component which has milk curdling activity. It has a broad range of substrate activity and demonstrates an esterase acitivity. It generally attacks peptide bonds.
TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) ∽ 35,000
pH Range 2.5-4.5 (2% aq soln)
Quantity 100 g
Source Porcine
Storage Requirements Room Temperature
Product Type Proteins, Enzymes & Peptides
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.